missing translation for 'onlineSavingsMsg'
Learn More

2,8-Dimethyldibenzothiophene, TRC

Product Code. 17914785
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17914785 250 mg 250mg
17910815 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17914785 Supplier Toronto Research Chemicals Supplier No. TRCD448280250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1207-15-4
Molecular Formula C14 H12 S
Molecular Weight (g/mol) 212.3101
IUPAC Name 2,8-dimethyldibenzothiophene
SMILES Cc1ccc2sc3ccc(C)cc3c2c1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H12S/c1-9-3-5-13-11(7-9)12-8-10(2)4-6-14(12)15-13/h3-8H,1-2H3
Formula Weight 212.066
Chemical Name or Material 2,8-Dimethyldibenzothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.