missing translation for 'onlineSavingsMsg'
Learn More

2-(9-Carbazolyl)ethylboronic acid pinacol ester, 98%

Product Code. 11335355
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335355 250 mg 250mg
11325355 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335355 Supplier Thermo Scientific Alfa Aesar Supplier No. L19580.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608534-41-4 | C20H24BNO2 | 321.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608534-41-4
Molecular Formula C20H24BNO2
Molecular Weight (g/mol) 321.227
MDL Number MFCD03788738
InChI Key NTUZDLWSQJRXAM-UHFFFAOYSA-N
Synonym 2-9-carbazolyl ethylboronic acid pinacol ester, 9-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl-9h-carbazole, 9-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl carbazole, 9-2-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl carbazole, 2-2-9h-carbazole-9-yl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 45925651
IUPAC Name 9-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)CCN2C3=CC=CC=C3C4=CC=CC=C42

Specifications

Melting Point 111°C to 113°C
Quantity 250 mg
Formula Weight 321.23
Percent Purity 98%
Chemical Name or Material 2-(9-Carbazolyl)ethylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.