missing translation for 'onlineSavingsMsg'
Learn More

2,9-Diacetylguanine, 96%, Thermo Scientific™

Product Code. 15560604 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15560604 25 g 25g
15570604 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15560604 Supplier Thermo Scientific Acros Supplier No. 459210250
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3056-33-5
Molecular Formula C9H9N5O3
Molecular Weight (g/mol) 235.20
MDL Number MFCD00142116
InChI Key GILZZWCROUGLIS-UHFFFAOYSA-N
PubChem CID 76461
IUPAC Name N-(9-acetyl-6-oxo-3H-purin-2-yl)acetamide
SMILES CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(C)=O

Specifications

Melting Point 285.0°C
Infrared Spectrum Authentic
Quantity 25 g
Synonym n2,9-diacetylguanine, n,9-diacetylguanine, n 2 ,9-diacetylguanine, 2,9-diacetylguanine, 2,9-diacetateguanine, n-9-acetyl-6-oxo-6,9-dihydro-1h-purin-2-yl acetamide, unii-vuw2b9yr95, 2,9-diacetyl guanine, vuw2b9yr95, n-9-acetyl-6-oxo-3h-purin-2-yl acetamide
Formula Weight 235.2
Percent Purity 96%
Chemical Name or Material 2,9-Diacetylguanine
compliance-icons
Product Identifier
  • 2,9-Diacetylguanine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.