missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dimethyl-1,10-phenanthroline, 98%

Product Code. 15435879
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435879 5 g 5g
15445879 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435879 Supplier Thermo Scientific Alfa Aesar Supplier No. J63698.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Reagent for the colorimetric determination of copper | CAS: 484-11-7 | C14H12N2 | 208.264 g/mol

2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

Solubility
Slightly soluble in water and light petroleum. Soluble in methanol, ethanol, acetone, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 484-11-7
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.264
MDL Number MFCD00004973
InChI Key IYRGXJIJGHOCFS-UHFFFAOYSA-N
Synonym neocuproine, neocuproin, 1,10-phenanthroline, 2,9-dimethyl, 2,9-dimethylphenanthroline, 2,9-dimethyl-o-phenanthroline, neo-cuproin, neocuproine hemihydrate, dmphen, unii-8e7d2sh3bv, 8e7d2sh3bv
PubChem CID 65237
IUPAC Name 2,9-dimethyl-1,10-phenanthroline
SMILES CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C

Spécification

Melting Point 159°C to 164°C
Color Brown to White
Quantity 5 g
Sensitivity Light sensitive
Merck Index 14,6449
Solubility Information Slightly soluble in water and light petroleum; Soluble in methanol,ethanol,acetone,ether and benzene.
Formula Weight 208.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,9-Dimethyl-1,10-phenanthroline
compliance-icons
Identifiant produit
  • 2,9-Dimethyl-1,10-phenanthroline
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Informations supplémentaires
  • MIXTURE LIST-Contains : 2,9-Dimethyl-1,10-phenanthroline

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.