missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetamido-2-deoxy-3-O-(Beta-D-galactopyranosyl)-D-galactose, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | 2-Acetamido-2-deoxy-3-O-(beta-D-galactopyranosyl)-D-galactose |
| CAS | 3554-90-3 |
| Molecular Formula | C14H25NO11 |
| Quantity | 5 mg |
| Synonym | beta-D-Galactosyl-(1-3)-N-acetylgalactosamine, Gal1-beta-3GalNAc, O-beta-D-Galactopyranosyl-(1-3)-N-acetylgalactosamine |
| InChI Formula | InChI=1S/C14H25NO11/c1-5(19)15-6(2-16)13(9(21)7(20)3-17)26-14-12(24)11(23)10(22)8(4-18)25-14/h2,6-14,17-18,20-24H,3-4H2,1H3,(H,15,19)/t6-,7+,8+,9-,10-,11-,12+,13+,14-/m0/s1 |
| SMILES | CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O |
| IUPAC Name | N-[(2R,3R,4S,5R)-4,5,6-trihydroxy-1-oxo-3-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexan-2-yl]acetamide |
| Molecular Weight (g/mol) | 383.34 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction