missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals

Product Code. 10687622 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10687622 100 g Plastic bottle 100g
10386160 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10687622 Supplier Thermo Scientific Acros Supplier No. 102561000
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10036-64-3
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00064359
InChI Key OVRNDRQMDRJTHS-PSLNIYNBNA-N
Synonym n-acetyl-alpha-d-glucosamine, alpha-glcnac, alpha-d-glcnac, 2-acetamido-2-deoxy-alpha-d-glucopyranose, unii-t13ti5gh3d, 2-acetylamino-2-deoxy-a-d-glucopyranose, t13ti5gh3d, 2-acetylamino-2-deoxy-alpha-d-glucopyranose, n-2s,3r,4r,5s,6r-2,4,5-trihydroxy-6-hydroxymethyl oxan-3-yl acetamide, n-2s,3r,4r,5s,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide
PubChem CID 82313
ChEBI CHEBI:44278
IUPAC Name N-[(2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1[C@@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White
Loss on Drying 1.0% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Quantity 100 g
Beilstein 31, 170
Merck Index 10, 4318
Specific Rotation Condition + 41.00 (20.00°C c=2,H2O)
Specific Rotation +39° to +43° (20°C, 589nm) (c=2, water)
Formula Weight 221.21
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material 2-Acetamido-2-deoxy-α-D-glucopyranose, 99.5+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.