missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Acetamido-2-deoxy-D-glucopyranose, 98%

Product Code. 10460521 Shop All Thermo Scientific Chemicals Products
Ansicht ändern
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Packungsgröße:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging Packungsgröße
10460521 10 g Glass bottle 10g
10696784 100 g Glass bottle 100g
10063863 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10460521 Lieferant Thermo Scientific Chemicals Lieferanten-Nr. 270940100
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
Synonym N-Acetyl-D-glucosamine
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Loss on Drying 2% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 4494
Specific Rotation Condition + 42.00 (20.00°C c=2,water,2 hrs)
Specific Rotation +39° to +45° (20°C, 589nm) (c=2, H2O, after 2 hrs)
Formula Weight 221.21
Percent Purity ≥97.5% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-2-deoxy-D-glucopyranose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.