missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-3-acetoxybenzoic Acid Methyl Ester, TRC

Product Code. 30326767
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326767 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326767 Supplier Toronto Research Chemicals Supplier No. TRCA16809525MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Acetamido-3-acetoxybenzoic Acid Methyl Ester
CAS 100193-10-0
Molecular Formula C12H13NO5
Quantity 25 mg
Synonym N-Acetyl-3-hydroxyanthranilic Acid Methyl Ester Acetate, 2-(Acetylamino)-3-(acetyloxy)benzoic Acid Methyl Ester
InChI Formula InChI=1S/C12H13NO5/c1-7(14)13-11-9(12(16)17-3)5-4-6-10(11)18-8(2)15/h4-6H,1-3H3,(H,13,14)
SMILES COC(=O)c1cccc(OC(=O)C)c1NC(=O)C
IUPAC Name methyl 2-acetamido-3-acetyloxybenzoate
Molecular Weight (g/mol) 251.24
Formula Weight 251.0794
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.