missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-3-O-(D-1-carboxyethyl)-2-deoxy-2-D-glucose, TRC

Product Code. 30179192
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179192 50 mg 50mg
30178957 25 mg 25mg
30179069 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179192 Supplier Toronto Research Chemicals Supplier No. TRCA14826550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10597-89-4
Molecular Formula C11H19NO8
Molecular Weight (g/mol) 293.27
Synonym N-Acetylmuramic Acid;
IUPAC Name (2R)-2-[(2R,3R,4R,5R)-2-acetamido-4,5,6-trihydroxy-1-oxohexan-3-yl]oxypropanoic acid
SMILES C[C@@H](O[C@@H]([C@H](O)[C@H](O)CO)[C@@H](NC(=O)C)C=O)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H19NO8/c1-5(11(18)19)20-10(9(17)8(16)4-14)7(3-13)12-6(2)15/h3,5,7-10,14,16-17H,4H2,1-2H3,(H,12,15)(H,18,19)/t5-,7+,8-,9-,10-/m1/s1
Formula Weight 293.1111
Chemical Name or Material 2-Acetamido-3-O-(D-1-carboxyethyl)-2-deoxy-2-D-glucose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.