missing translation for 'onlineSavingsMsg'
Learn More

2-Acetoacetotoluidide, 99+%, Thermo Scientific™

Product Code. 11317855
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317855 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11317855 Supplier Thermo Scientific Alfa Aesar Supplier No. L02812.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 93-68-5
Melting Point 104°C to 106°C
Density 1.062
Flash Point 143°C (289°F)
Odor Odorless
Molecular Formula C11H13NO2
MDL Number MFCD00008782
Quantity 100 g
Beilstein 2099098
Synonym 2'-methylacetoacetanilide, n-2-methylphenyl-3-oxobutanamide, o-acetoacetotoluidide, n-acetoacetyl-o-toluidine, acetoacet-o-toluidide, acetoacet-o-toluidine, 2-acetoacetylaminotoluene, 3-oxo-n-o-tolyl butanamide, butanamide, n-2-methylphenyl-3-oxo, acetoacetyl-2-methylanilide
InChI Key TVZIWRMELPWPPR-UHFFFAOYSA-N
SMILES CC(=O)CC(=O)NC1=CC=CC=C1C
IUPAC Name N-(2-methylphenyl)-3-oxobutanamide
Molecular Weight (g/mol) 191.23
PubChem CID 7154
Formula Weight 191.23
Percent Purity ≥99%
Chemical Name or Material 2-Acetoacetotoluidide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.