missing translation for 'onlineSavingsMsg'
Learn More

2-Acetoxyisobutyryl bromide, 96%

Product Code. 11446780
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446780 100 g 100g
11426780 5 g 5g
11436780 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446780 Supplier Thermo Scientific Alfa Aesar Supplier No. L11592.22
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40635-67-4 | C6H9BrO3 | 209.039 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40635-67-4
Molecular Formula C6H9BrO3
Molecular Weight (g/mol) 209.039
MDL Number MFCD00040923
InChI Key OOKAXSHFTDPZHP-UHFFFAOYSA-N
Synonym 2-acetoxyisobutyryl bromide, 2-acetoxy-2-methylpropionyl bromide, 1-bromocarbonyl-1-methylethyl acetate, alpha-acetoxy-isobutyryl bromide, a-acetoxyisobutyryl bromide, alpha-acetoxyisobutyryl bromide, 2-acetoxyisobutyryl bromide;2-acetoxy-2-methylpropionyl bromide, propanoyl bromide, 2-acetyloxy-2-methyl, aibb, pubchem17437
PubChem CID 2724715
IUPAC Name (1-bromo-2-methyl-1-oxopropan-2-yl) acetate
SMILES CC(=O)OC(C)(C)C(=O)Br

Specifications

Density 1.42
Boiling Point 100°C to 110°C (25 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.457
Quantity 100 g
UN Number UN3265
Beilstein 2432585
Sensitivity Moisture sensitive
Formula Weight 209.05
Percent Purity 96%
Chemical Name or Material 2-Acetoxyisobutyryl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.