missing translation for 'onlineSavingsMsg'
Learn More

2-(Acetoxymethoxy)-1,3-propanediyl Dibenzoate, TRC

Product Code. 30322442
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322442 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322442 Supplier Toronto Research Chemicals Supplier No. TRCA166570100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Acetoxymethoxy)-1,3-propanediyl Dibenzoate
CAS 110874-22-1
Molecular Formula C20 H20 O7
Quantity 100 mg
InChI Formula InChI=1S/C20H20O7/c1-15(21)26-14-27-18(12-24-19(22)16-8-4-2-5-9-16)13-25-20(23)17-10-6-3-7-11-17/h2-11,18H,12-14H2,1H3
SMILES CC(=O)OCOC(COC(=O)c1ccccc1)COC(=O)c2ccccc2
IUPAC Name [2-(acetyloxymethoxy)-3-benzoyloxypropyl] benzoate
Molecular Weight (g/mol) 372.37
Formula Weight 372.1209
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.