missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-3-aminothiophene, 97%

Product Code. 11388505
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388505 250 mg 250mg
11378505 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388505 Supplier Thermo Scientific Alfa Aesar Supplier No. L10563.MD
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31968-33-9 | C6H7NOS | 141.188 g/mol

Dithienopyridines was synthesised from 2-Acetyl-3-aminothiophene. Also used as fine Intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dithienopyridines was synthesised from 2-Acetyl-3-aminothiophene. Also used as fine Intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31968-33-9
Molecular Formula C6H7NOS
Molecular Weight (g/mol) 141.188
MDL Number MFCD00052585
InChI Key MMLSKDGCLMDHAJ-UHFFFAOYSA-N
Synonym 2-acetyl-3-aminothiophene, 1-3-aminothiophen-2-yl ethanone, ethanone, 1-3-amino-2-thienyl, ethanone,1-3-amino-2-thienyl, pubchem20830, acmc-1co94, 1-3-aminothien-2-yl ethanone, 2-acetylthiophene-3-amine, 1-3-amino-2-thiophenyl ethanone
PubChem CID 2735233
IUPAC Name 1-(3-aminothiophen-2-yl)ethanone
SMILES CC(=O)C1=C(C=CS1)N

Specifications

Melting Point 84°C to 88°C
Boiling Point 120°C to 125°C (0.8 mmHg)
Quantity 250 mg
Beilstein 1635973
Solubility Information Insoluble in water.
Formula Weight 141.19
Percent Purity 97%
Chemical Name or Material 2-Acetyl-3-aminothiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.