missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole, 97%

Product Code. 15498296
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 15498296

Brand: Thermo Scientific Alfa Aesar H63416.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94944-70-4 | C9H14N2O5 | 230.22 g/mol

2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole used for treatment of autoimmune disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole used for treatment of autoimmune disorders.

Solubility
Soluble in distilled water, DMSO (>5 mg/ml), clear (warmed), and PBS pH 7.2 (∼0.15 mg/ml).

Notes
TRUSTED_SUSTAINABILITY

Specifications

CAS 94944-70-4
Color White to Brown
Molecular Formula C9H14N2O5
MDL Number MFCD08752516
Quantity 250 mg
Synonym 2-athbi, 2-acetyl-4 5-tetrahydroxybutylimidazole, unii-hw195j55g1, thi, 2-acetyl-5-tetrahydroxybutyl imidazole, 2-acetyl-4-tetrahydroxybutylimidazole, ethanone, 1-4-1,2,3,4-tetrahydroxybutyl-1h-imidazol-2-yl-, 1r-1r*,2s*,3r*, 2-acetyl-4 5-1,2,3,4-tetrahydroxybutyl-imidazole thi, 1-5-1r,2s,3r-1,2,3,4-tetrahydroxybutyl-1h-imidazol-2-yl ethanone, 2-acetyl-4-tetrahydroxybutyl imidazole; 1-5-1r,2s,3r-1,2,3,4-tetrahydroxybutyl-1h-imidazol-2-yl-ethanone
Solubility Information Soluble in distilled water,DMSO (>5mg/ml),clear (warmed),and PBS pH 7.2 (∽0.15mg/ml).
InChI Key CQSIXFHVGKMLGQ-BWZBUEFSSA-N
SMILES CC(=O)C1=NC=C(N1)C(C(C(CO)O)O)O
IUPAC Name 1-[5-[(1R,2S,3R)-1,2,3,4-tetrahydroxybutyl]-1H-imidazol-2-yl]ethanone
Molecular Weight (g/mol) 230.22
PubChem CID 108037
Formula Weight 230.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.