missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-chlorothiophene, 99%, Thermo Scientific™

Product Code. 10023690 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10023690

Brand: Thermo Scientific Acros 306700050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6310-09-4
Molecular Weight (g/mol) 160.62
MDL Number MFCD00005444
InChI Key HTZGPEHWQCRXGZ-UHFFFAOYSA-N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 43°C to 47°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H5ClOS
Quantity 5 g
Synonym 2-acetyl-5-chlorothiophene, 1-5-chlorothiophen-2-yl ethanone, 1-5-chloro-2-thienyl ethanone, 5-chloro-2-acetylthiophene, ethanone, 1-5-chloro-2-thienyl, 5-chlor-2-acetyl thiophen, ketone, 5-chloro-2-thienyl methyl, 1-5-chlorothiophen-2-yl ethan-1-one, 5-chloro-2-thienyl methyl ketone, 2-chloro-5-acetylthiophene
SMILES CC(=O)C1=CC=C(S1)Cl
IUPAC Name 1-(5-chlorothiophen-2-yl)ethanone
PubChem CID 80572
Formula Weight 160.62
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Acetyl-5-chlorothiophene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.