missing translation for 'onlineSavingsMsg'
Learn More

2-(Acetylamino)-N-(phenylmethyl)-2-propenamide, TRC

Product Code. 30320191
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320191 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320191 Supplier Toronto Research Chemicals Supplier No. TRCA16435025MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Acetamido-N-benzylprop-2-enamide
CAS 86921-49-5
Purity Grade Notes HPLC
Molecular Formula C12 H14 N2 O2
Quantity 25 mg
Synonym 2-Propenamide, 2-(acetylamino)-N-(phenylmethyl)-, 2-(Acetylamino)-N-(phenylmethyl)-2-propenamide, 2-Acetamido-N-benzylacrylamide, 2-(Acetylamino)-N-benzylprop-2-enamide, 2-Acetamido-N-benzylprop-2-enamide, Lacosamide Imp. K (EP)
InChI Formula InChI=1S/C12H14N2O2/c1-9(14-10(2)15)12(16)13-8-11-6-4-3-5-7-11/h3-7H,1,8H2,2H3,(H,13,16)(H,14,15)
SMILES CC(=O)NC(=C)C(=O)NCc1ccccc1
IUPAC Name 2-acetamido-N-benzylprop-2-enamide
Molecular Weight (g/mol) 218.2518
Formula Weight 218.1055
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.