missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbutyrolactone, 99+%

Product Code. 10520604 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
Unit Size:
1 L
100mL
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10520604 1 L 1 L
10745551 100 mL 100mL
10438050 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10520604 Supplier Thermo Scientific Acros Supplier No. 102490010
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 517-23-7 | C6H8O3 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-23-7
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00005394
InChI Key OMQHDIHZSDEIFH-UHFFFAOYNA-N
Synonym 2-acetylbutyrolactone, 3-acetyldihydrofuran-2 3h-one, alpha-acetylbutyrolactone, alpha-acetyl-gamma-butyrolactone, 3-acetyldihydro-2 3h-furanone, 2 3h-furanone, 3-acetyldihydro, 3-acetyltetrahydrofuran-2-one, 2-oxo-3-acetyltetrahydrofuran, 3-acetyltetrahydro-2-furanone, 3-acetyl-2 3h-4,5-dihydrofuranone
PubChem CID 10601
IUPAC Name 3-acetyloxolan-2-one
SMILES CC(=O)C1CCOC1=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Density 1.1800g/mL
Boiling Point 107°C to 108°C (5.0 mmHg)
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4585
Quantity 1 L
Merck Index 15, 78
Specific Gravity 1.18
Solubility Information Solubility in water: 310g/L (20°C)
Formula Weight 128.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Acetylbutyrolactone, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Acetylbutyrolactone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.