missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyridine, 98%

Product Code. 11966281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11966281 500 g Glass bottle 500g
10408630 25 g Glass bottle 25g
10346260 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11966281 Supplier Thermo Scientific Acros Supplier No. 102685000
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1122-62-9 | C7H7NO | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-62-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
InChI Key AJKVQEKCUACUMD-UHFFFAOYSA-N
Synonym 2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone
PubChem CID 14286
IUPAC Name 1-pyridin-2-ylethanone
SMILES CC(=O)C1=CC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 8.0°C to 10.0°C
Color Colorless to Brown
Density 1.0800g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C5H4NCOCH3
Refractive Index 1.5200 to 1.5220
Quantity 500 g
Specific Gravity 1.08
Solubility Information Solubility in water: 200g/L (20°C). Other solubilities: miscible with ethanol
Formula Weight 121.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Acetylpyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Acetylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.