missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyrrole, 98%

Product Code. 11412018
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412018 25 g 25g
11492008 1 g 1g
11402018 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412018 Supplier Thermo Scientific Alfa Aesar Supplier No. A14593.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1072-83-9 | C6H7NO | 109.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-83-9
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00005220
InChI Key IGJQUJNPMOYEJY-UHFFFAOYSA-N
Synonym 2-acetylpyrrole, 1-1h-pyrrol-2-yl ethanone, 1-1h-pyrrol-2-yl ethan-1-one, 2-acetyl pyrrole, 2-pyrrolylethanone, 2-acetyl-1h-pyrrole, methyl pyrrol-2-yl ketone, ethanone, 1-1h-pyrrol-2-yl, 2-pyrrolyl methyl ketone, ketone, methyl pyrrol-2-yl
PubChem CID 14079
ChEBI CHEBI:59981
IUPAC Name 1-(1H-pyrrol-2-yl)ethanone
SMILES CC(=O)C1=CC=CN1

Specifications

Melting Point 88°C to 92°C
Boiling Point 220°C
Odor Characteristic
Quantity 25 g
Beilstein 1882
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2-Acetylpyrrole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.