missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylquinoxaline, 97%, Thermo Scientific Chemicals

Product Code. 11352609
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352609 1 g 1g
15403527 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352609 Supplier Thermo Scientific Alfa Aesar Supplier No. H26680.03
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

2-Acetylquinoxaline is used as dyes, pharmaceuticals, and antibiotics such as olaquindox, carbadox, echinomycin, levomycin and actinoleutin. It is also used to make amiloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylquinoxaline is used as dyes, pharmaceuticals, and antibiotics such as olaquindox, carbadox, echinomycin, levomycin and actinoleutin. It is also used to make amiloride.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25594-62-1
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD07808559
InChI Key SWGILLQQUULMJB-UHFFFAOYSA-N
Synonym 2-acetylquinoxaline, 1-quinoxalin-2-yl ethanone, ethanone, 1-2-quinoxalinyl, acmc-1cknl, 1-quinoxalin-2-yl ethan-1-one
PubChem CID 11105814
IUPAC Name 1-quinoxalin-2-ylethanone
SMILES CC(=O)C1=NC2=CC=CC=C2N=C1

Specifications

Melting Point 76°C to 79°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 2-Acetylquinoxaline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.