missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanone, 98%

Product Code. 11482427
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482427 5 g 5g
11492427 25 g 25g
11402437 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482427 Supplier Thermo Scientific Alfa Aesar Supplier No. A14275.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 700-58-3 | C10H14O | 150.221 g/mol

It is used in the synthesis of dispiro N-Boc-protected 1,2,4-trioxane1 and (±)-1-(adamantan-2-yl)-2-propanamine. It is employed in reductive coupling (TiCl3/Li) which gives (adamantylidene)adamantane, an example of a general method for the synthesis of highly-substituted alkenes and in the preparation of highly-substituted alkenes by the Wittig reaction which gives poor yields with adamantanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of dispiro N-Boc-protected 1,2,4-trioxane1 and (+/-)-1-(adamantan-2-yl)-2-propanamine. It is employed in reductive coupling (TiCl3/Li) which gives (adamantylidene)adamantane, an example of a general method for the synthesis of highly-substituted alkenes and in the preparation of highly-substituted alkenes by the Wittig reaction which gives poor yields with adamantanone.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-58-3
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00074737
InChI Key IYKFYARMMIESOX-UHFFFAOYSA-N
Synonym 2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
PubChem CID 64151
ChEBI CHEBI:40611
IUPAC Name adamantan-2-one
SMILES C1C2CC3CC1CC(C2)C3=O

Specifications

Melting Point ∼257°C (sublimation)
Quantity 5 g
Beilstein 1210235
Formula Weight 150.22
Percent Purity 98%
Chemical Name or Material 2-Adamantanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.