missing translation for 'onlineSavingsMsg'
Learn More

2-(Alpha-D-Mannopyranosyl)-L-tryptophan, TRC

Product Code. 30136381
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136381 10 mg 10mg
30004851 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136381 Supplier Toronto Research Chemicals Supplier No. TRCM16645010MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180509-18-6
Molecular Formula C17H22N2O7
Molecular Weight (g/mol) 366.37
Synonym C2-α-D-Mannopyranosyltryptophan, 2-α-D-Mannopyranosyl-L-tryptophan, 2-α-D-Mannopyranosyl-L-tryptophan
IUPAC Name (S)-2-amino-3-(2-((2R,3S,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=C([C@@H]2[C@H]([C@H]([C@@H]([C@@H](CO)O2)O)O)O)NC3=C1C=CC=C3)C(O)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H22N2O7/c18-9(17(24)25)5-8-7-3-1-2-4-10(7)19-12(8)16-15(23)14(22)13(21)11(6-20)26-16/h1-4,9,11,13-16,19-23H,5-6,18H2,(H,24,25)/t9-,11+,13+,14-,15-,16+/m0/s1
Formula Weight 366.14
Percent Purity >95%
Chemical Name or Material 2-(alpha-D-Mannopyranosyl)-L-tryptophan
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.