missing translation for 'onlineSavingsMsg'
Learn More

2-Amidinopyridine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 13555010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13555010 5 g 5g
13565010 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13555010 Supplier Thermo Scientific Acros Supplier No. 448610050
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51285-26-8 | Molecular Formula: C6H7N3·ClH | Molecular Weight: 157.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51285-26-8
Molecular Formula C6H7N3·ClH
Molecular Weight (g/mol) 157.6
InChI Key GMHCEDDZKAYPLB-UHFFFAOYSA-N
Synonym picolinimidamide hydrochloride, pyridine-2-carboximidamide hydrochloride, 2-amidinopyridine hydrochloride, pyridine-2-carboxamidine hydrochloride, picolinamidine hydrochloride, picolinamidine hcl, picolinimidamide hcl, pyridine-2-carboximidamide, hcl, 2-amidinopyridine, 2-amidinopyridinium chloride
PubChem CID 12403615
IUPAC Name pyridine-2-carboximidamide;hydrochloride
SMILES C1=CC=NC(=C1)C(=N)N.Cl

Specifications

Melting Point 146.0°C to 148.0°C
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 157.6
Percent Purity 97%
Chemical Name or Material 2-Amidinopyridine Hydrochloride
compliance-icons
Product Identifier
  • 2-Amidinopyridine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.