missing translation for 'onlineSavingsMsg'
Learn More

(±)-2-Amino-1-butanol, 97%

Product Code. 10366160 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366160 100 mL Glass bottle 100mL
10588460 5 mL Glass bottle 5mL
10102370 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10366160 Supplier Thermo Scientific Acros Supplier No. 103251000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 96-20-8 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-20-8
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00008095
InChI Key JCBPETKZIGVZRE-UHFFFAOYSA-N
Synonym 2-amino-1-butanol, 2-aminobutanol, dl-2-amino-1-butanol, 1-butanol, 2-amino, 2-aminobutyl alcohol, 1-hydroxy-2-butylamine, butanol-2-amine, 1-hydroxymethyl propylamine, 2-amino-1-hydroxybutane, 2-amino-n-butyl alcohol
PubChem CID 22129
IUPAC Name 2-aminobutan-1-ol
SMILES CCC(CO)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -2°C
Density 0.9440g/mL
Boiling Point 176°C to 178°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(NH2)CH2OH
Refractive Index 1.4508 to 1.4528
Quantity 100 mL
Merck Index 15, 422
Specific Gravity 0.944
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and ether
Formula Weight 89.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (±)-2-Amino-1-butanol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (+/-)-2-Amino-1-butanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.