missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Amino-1-butanol, 97%

Product Code. 11435847
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435847 100 mL 100mL
11445847 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435847 Supplier Thermo Scientific Alfa Aesar Supplier No. B24398.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 96-20-8 | C4H11NO | 89.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-20-8
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00008095
InChI Key JCBPETKZIGVZRE-UHFFFAOYSA-N
Synonym 2-amino-1-butanol, 2-aminobutanol, dl-2-amino-1-butanol, 1-butanol, 2-amino, 2-aminobutyl alcohol, 1-hydroxy-2-butylamine, butanol-2-amine, 1-hydroxymethyl propylamine, 2-amino-1-hydroxybutane, 2-amino-n-butyl alcohol
PubChem CID 22129
IUPAC Name 2-aminobutan-1-ol
SMILES CCC(CO)N

Specifications

Melting Point -2°C
Density 0.944
pH 11.1
Boiling Point 176°C to 178°C
Flash Point 89°C (192°F)
Odor Amine-like
Refractive Index 1.4525
Quantity 100 mL
UN Number UN2735
Beilstein 1098274
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,427
Formula Weight 89.14
Percent Purity 97%
Chemical Name or Material (+/-)-2-Amino-1-butanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (+/-)-2-Amino-1-butanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Amino-1-butanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.