missing translation for 'onlineSavingsMsg'
Learn More

(±)-2-Amino-1-phenylethanol, 95%

Product Code. 10436953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10436953 10 g Glass bottle 10g
10302583 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10436953 Supplier Thermo Scientific Acros Supplier No. 104320100
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7568-93-6 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7568-93-6
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008137
InChI Key ULSIYEODSMZIPX-UHFFFAOYNA-N
Synonym phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanol
PubChem CID 1000
ChEBI CHEBI:16343
IUPAC Name 2-amino-1-phenylethanol
SMILES NCC(O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 56.0°C to 60.0°C
Color Yellow
Boiling Point 160.0°C (17.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NCH2CH(C6H5)OH
Refractive Index 1.5580 to 1.5600
Quantity 10 g
Merck Index 15, 7399
Formula Weight 137.18
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material (±)-2-Amino-1-phenylethanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (#1)-2-Amino-1-phenylethanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.