missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-cyanoacetamide, 97+%

Product Code. 10270103 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10270103 25 g Glass bottle 25g
10168152 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10270103 Supplier Thermo Scientific Acros Supplier No. 382760250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6719-21-7 | C3H5N3O | 99.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6719-21-7
Molecular Formula C3H5N3O
Molecular Weight (g/mol) 99.09
MDL Number MFCD00040532
InChI Key JRWAUKYINYWSTA-UHFFFAOYSA-N
Synonym aminocyanoacetamide, 3-nitriloalaninamide, acetamide, 2-amino-2-cyano, 2-aminocyanoacetamide, aminocyanacetamide, 2-amino-cyanoacetamide, 2-amino-2cyanoacetamide, a-amino-a-cyanoacetamide, 2-amino-2-cyanacetamide, 2-amino 2-cyano acetamide
PubChem CID 98341
IUPAC Name 2-amino-2-cyanoacetamide
SMILES C(#N)C(C(=O)N)N

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 120°C to 124°C
Color Beige to Tan
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula NCCH(NH2)CONH2
Quantity 25 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ether and methanol
Formula Weight 99.09
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 2-Amino-2-cyanoacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-2-cyanoacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.