missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Amino-2'-deoxyadenosine, 99%

Product Code. 15466289
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466289 1 g 1g
15476289 5 g 5g
15486289 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15466289 Supplier Thermo Scientific Chemicals Supplier No. J64266.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4546-70-7 | C10H14N6O3 | 266.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4546-70-7
Molecular Formula C10H14N6O3
Molecular Weight (g/mol) 266.26
MDL Number MFCD00047240
InChI Key NOLHIMIFXOBLFF-DWFKHXNSNA-N
Synonym 2-amino-2'-deoxyadenosine, 2,6-diaminopurine-2'-deoxyriboside, adenosine, 2-amino-2'-deoxy, 2,6-diaminopurinedeoxyriboside, 2,6-diaminopurine 2'-deoxyriboside, unii-4zph2f1jhr, 4zph2f1jhr, 9h-purine, 9-2-deoxy-beta-d-erythro-pentofuranosyl-2,6-diamino, 2-amino-deoxyadenosine, 2-amino-2/'-deoxyadenosine
PubChem CID 97188
IUPAC Name (2R,3S,5R)-5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
SMILES NC1=NC(N)=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=N1

Specifications

Color White
Quantity 1 g
Formula Weight 266.26
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Amino-2'-deoxyadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.