missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Amino-2'-deoxyguanosine, 98%

Product Code. 15436499
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436499 250 mg 250mg
15426499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436499 Supplier Thermo Scientific Chemicals Supplier No. J64650.MD
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60966-26-9 | C10H14N6O4 | 282.26 g/mol

2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60966-26-9
Molecular Formula C10H14N6O4
Molecular Weight (g/mol) 282.26
MDL Number MFCD01723954
InChI Key ROPTVRLUGSPXNH-SJLGBTOWNA-N
PubChem CID 173775
IUPAC Name 2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O

Specifications

Color White
Quantity 250 mg
Synonym 2'-amino-2'-deoxyguanosine, guanosine, 2'-amino-2'-deoxy, 2'-deoxy-2'-aminoguanosine, 2ag, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one, guanosine,2'-amino-2'-deoxy, 2'-amino-2'-deoxyguanosine,2'-nh2-dg, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one
Formula Weight 282.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2'-Amino-2'-deoxyguanosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.