missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-(hydroxymethyl)propane-1,3-diol 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate, 98%, BLD Pharm™

Product Code. 31431045
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31431045 5 g
31427273 1 g
31417766 100 mg
31417881 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31431045 Supplier BLD Pharm Supplier No. BD1355855g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 74103-07-4
Molecular Formula C19H24N2O6
MDL Number MFCD00887595
Quantity 5 g
SMILES O=C(C1C2=CC=C(C(C3=CC=CC=C3)=O)N2CC1)O.OCC(CO)(N)CO
Molecular Weight (g/mol) 376.40
Percent Purity 98%
Chemical Name or Material 2-Amino-2-(hydroxymethyl)propane-1,3-diol 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.