missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1-propanol, 95%, may cont. ca 5% water

Product Code. 11453408
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453408 500 mL 500mL
11349699 100 mL 100mL
11463408 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453408 Supplier Thermo Scientific Alfa Aesar Supplier No. A17814.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124-68-5 | C4H11NO | 89.14 g/mol

2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.

Solubility
Immiscible with water. Miscible with alcohols.

Notes
Incompatible withoxidizing agents, strong acids, copper, brass and aluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-68-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00008051
InChI Key CBTVGIZVANVGBH-UHFFFAOYSA-N
Synonym 2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
PubChem CID 11807
IUPAC Name 2-amino-2-methylpropan-1-ol
SMILES CC(C)(N)CO

Specifications

Melting Point 24°C to 28°C
Density 0.934
pH 11.3
Boiling Point 164°C to 166°C
Flash Point 67°C (152°F)
Odor Amine-like
Refractive Index 1.4455
Quantity 500 mL
Beilstein 505979
Merck Index 14,449
Solubility Information Immiscible with water. Miscible with alcohols.
Formula Weight 89.14
Percent Purity 95%
Assay may cont. ≈5% water
Chemical Name or Material 2-Amino-2-methyl-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-2-methyl-1-propanol, 95%, may contain ca 5% water
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Amino-2-methyl-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.