missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1-propanol, 99%

Product Code. 10724211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10724211 1 L Glass bottle 1 L
10615712 500 mL Glass bottle 500mL
10404025 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10724211 Supplier Thermo Scientific Acros Supplier No. 104060010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124-68-5 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-68-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00008051
InChI Key CBTVGIZVANVGBH-UHFFFAOYSA-N
Synonym 2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
PubChem CID 11807
IUPAC Name 2-amino-2-methylpropan-1-ol
SMILES CC(C)(N)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 31.0°C to 32.0°C
Color Colorless
Density 0.9300g/mL
Boiling Point 165.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2C(NH2)CH2OH
Refractive Index 1.4470 to 1.4490
Quantity 1 L
Beilstein 04, III, 783
Fieser 01,37; 03,14; 06,20
Merck Index 15, 446
Specific Gravity 0.93
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohols
Viscosity 102 mPa.s (30°C)
Formula Weight 89.14
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 2-Amino-2-methyl-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-2-methyl-1-propanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-Amino-2-methyl-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.