missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-Methyl-Propan-1-ol, Extra Pure, SLR, Fisher Chemical

Product Code. 10020030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10020030 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10020030 Supplier Fisher Chemical Supplier No. A/2960/08
  • ONLINE EXCLUSIVE PRICE
    574.00 €
    568.00 € / 500mL
    Save 6.00 €
    1% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 124-68-5
Melting Point 31°C
Color Colorless
pH 11.3
Boiling Point 165°C
Packaging Amber glass bottle
Molecular Formula C4H11NO
MDL Number 8051
Quantity 500 mL
Formula Weight 89.14g/mol
Physical Form Low Melting Solid, Solid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-2-methyl-1-propanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.