missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methylpropanol (~95%), TRC

Product Code. 30318655
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318655 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318655 Supplier Toronto Research Chemicals Supplier No. TRCA61740010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-2-methylpropanol
CAS 124-68-5
Molecular Formula C4 H11 N O
Quantity 10 g
Synonym 2-Amino-2-methyl-1-propanol, 1,1-Dimethyl-2-hydroxyethylamine, 1-Hydroxy-2-methylpropan-2-amine, 2,2-Dimethylethanolamine, 2-Amino-1-hydroxy-2-methylpropane, 2-Amino-2,2-dimethylethanol, 2-Amino-2-methylpropanol, 2-Aminoisobutanol, 2-Hydroxy-1,1-dimethylethylamine, 2-Hydroxymethyl-2-propylamine, 2-Methyl-2-amino-1-propanol, 2-Methyl-2-aminopropanol, AMP, AMP (thinner), AMP 100, AMP 75, AMP 90, AMP 90 (amine), AMP 95, AMP Regular, AMP buffer, AMP-ULTRA PC 2000, AMP-Ultra PC, AMP-Ultra PC 1000, CE 64, Corrguard 75, Dolapix A 88, Hydroxy-tert-butylamine, Isobutanol-2-amine, KV 5088, NSC 441, Pamabron, β-Aminoisobutanol
InChI Formula InChI=1S/C4H11NO/c1-4(2,5)3-6/h6H,3,5H2,1-2H3
SMILES CC(C)(N)CO
IUPAC Name 2-amino-2-methylpropan-1-ol
Molecular Weight (g/mol) 89.14
Formula Weight 89.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.