missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromopyrazine, 97%, Thermo Scientific™

Product Code. 10091394 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10091394 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10091394 Supplier Thermo Scientific Acros Supplier No. 398370010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 24241-18-7
CAS Min % 96.0
CAS Max % 100.0
Melting Point 114°C to 118°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C4H3Br2N3
MDL Number MFCD00673150
Quantity 1 g
Synonym 2-amino-3,5-dibromopyrazine, 3,5-dibromo-2-pyrazinamine, 3,5-dibromopyrazin-2-ylamine, 3,5-dibromopyrazine-2-ylamine, amino-3,5-dibromopyrazine, pyrazinamine, 3,5-dibromo, 3,5-dibromopyrazine-2-amine, 2-amino-3,5-dibromo pyrazine, 2-amino-3,5-dibromo-pyrazine, 2-pyrazinamine, 3,5-dibromo
InChI Key DTLBKXRFWUERQN-UHFFFAOYSA-N
SMILES NC1=NC=C(Br)N=C1Br
IUPAC Name 3,5-dibromopyrazin-2-amine
Molecular Weight (g/mol) 252.90
PubChem CID 620004
Formula Weight 252.89
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 2-Amino-3, 5-dibromopyrazine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3,5-dibromopyrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.