missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-fluoropyridine, 97%, Thermo Scientific Chemicals

Product Code. 15457747
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457747 5 g 5g
15447747 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457747 Supplier Thermo Scientific Alfa Aesar Supplier No. H37257.06
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869557-43-7
Molecular Formula C5H4BrFN2
Molecular Weight (g/mol) 191.00
MDL Number MFCD03092924
InChI Key KXSQMCRVUAALNE-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-fluoropyridine, 2-amino-5-fluoro-3-bromopyridine, 3-bromo-5-fluoro-pyridin-2-ylamine, 3-bromo-5-fluoro-2-pyridinamine, 2-pyridinamine, 3-bromo-5-fluoro, 3-bromo-5-fluoro-2-pyridylamine, pubchem1101, acmc-209qcf, ksc490s4d, 3-bromo-5-fluoro-2-pyridineamine
PubChem CID 16126434
IUPAC Name 3-bromo-5-fluoropyridin-2-amine
SMILES NC1=C(Br)C=C(F)C=N1

Specifications

Boiling Point 63°C to 65°C
Quantity 5 g
Formula Weight 191
Percent Purity 97%
Chemical Name or Material 2-Amino-3-bromo-5-fluoropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.