missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methyl-3H-imidazo[4,5-f]quinoxaline, TRC

Product Code. 17961375
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17961375 2.5 mg 2.5mg
17904295 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17961375 Supplier Toronto Research Chemicals Supplier No. TRCA6169002.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108354-47-8
Molecular Formula C10 H9 N5
Molecular Weight (g/mol) 199.21
Synonym 3-Methyl-3H-imidazo[4,5-f]quinoxalin-2-amine, 2-Amino-3-Methylimidazo[4,5-f]quinoxaline, 3-Methylimidazo[4,5-f]quinoxalin-2-amine
IUPAC Name 3-methylimidazo[4,5-f]quinoxalin-2-amine
SMILES Cn1c(N)nc2c3nccnc3ccc12

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C10H9N5/c1-15-7-3-2-6-8(13-5-4-12-6)9(7)14-10(15)11/h2-5H,1H3,(H2,11,14)
Formula Weight 199.0858
Percent Purity >95%
Chemical Name or Material 2-Amino-3-methyl-3H-imidazo[4,5-F]quinoxaline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.