missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methylbenzoic Acid, TRC

Product Code. 30316564
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316564 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316564 Supplier Toronto Research Chemicals Supplier No. TRCA6121505G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-3-methylbenzoic Acid
CAS 4389-45-1
Molecular Formula C8H9NO2
Quantity 5 g
Synonym 2-Amino-3-methylbenzoic Acid, 2-Amino-m-toluic Acid, 3-Methyl-2-aminobenzoic Acid, 3-Methylanthranilic Acid, NSC 16049
InChI Formula InChI=1S/C8H9NO2/c1-5-3-2-4-6(7(5)9)8(10)11/h2-4H,9H2,1H3,(H,10,11)
SMILES Cc1cccc(C(=O)O)c1N
IUPAC Name 2-amino-3-methylbenzoic acid
Molecular Weight (g/mol) 151.16
Formula Weight 151.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.