missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,4,4-trifluorobutyric acid, 97%

Product Code. 11367916
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367916 100 mg 100mg
11357916 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367916 Supplier Thermo Scientific Alfa Aesar Supplier No. L13131.MC
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15959-93-0 | C4H6F3NO2 | 157.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15959-93-0
Molecular Formula C4H6F3NO2
Molecular Weight (g/mol) 157.092
MDL Number MFCD00041417
InChI Key AQPCXCOPDSEKQT-UHFFFAOYSA-N
PubChem CID 353083
IUPAC Name 2-amino-4,4,4-trifluorobutanoic acid
SMILES C(C(C(=O)O)N)C(F)(F)F

Specifications

Melting Point ∽250°C (decomposition)
Quantity 100 mg
Beilstein 1769355
Synonym 2-amino-4,4,4-trifluorobutyric acid, butanoic acid, 2-amino-4,4,4-trifluoro, s-2-amino-4,4,4-trifluorobutyric acid, 3-trifluoromethylalanine, 2-amino-4,4,4-trifluoro-butyric acid, butanoicacid,2-amino-4,4,4-trifluoro, butanoic acid,2-amino-4,4,4-trifluoro, 2-amino-4,4,4-trifluoro-n-butyric acid hydrochlori
Formula Weight 157.09
Percent Purity 97%
Chemical Name or Material 2-Amino-4,4,4-trifluorobutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.