missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(4-cyanophenyl)thiazole, 97%

Product Code. 15471028
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15471028 250 mg 250mg
15451028 1 g 1g
15461028 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471028 Supplier Thermo Scientific Alfa Aesar Supplier No. H54861.MD
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 436151-85-8 | C10H7N3S | 201.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 436151-85-8
Molecular Formula C10H7N3S
Molecular Weight (g/mol) 201.25
MDL Number MFCD03407828
InChI Key KKMZKOIZTSRIEM-UHFFFAOYSA-N
Synonym 4-2-amino-1,3-thiazol-4-yl benzonitrile, 4-2-aminothiazol-4-yl benzonitrile, 4-2-amino-thiazol-4-yl-benzonitrile, benzonitrile, 4-2-amino-4-thiazolyl, acmc-1aqww, 2-amino-4-4-cyanophenyl thiazole, 4-4-cyanophenyl-2-aminothiazole, 4-2-amino-4-thiazolyl benzonitrile, 4-2-aminothiazole-4-yl benzonitrile, 4-2-amino-thiazol-4-yl benzonitrile
PubChem CID 2049589
IUPAC Name 4-(2-amino-1,3-thiazol-4-yl)benzonitrile
SMILES NC1=NC(=CS1)C1=CC=C(C=C1)C#N

Specifications

Quantity 250 mg
Beilstein 15029305
Formula Weight 201.25
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(4-cyanophenyl)thiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.