missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-4-(4-methylphenyl)pyrimidine, 96%

Product Code. 11459540
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459540 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11459540 Supplier Alfa Aesar™ Supplier No. H50357.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 263276-44-4
Molecular Formula C11H11N3
Molecular Weight (g/mol) 185.23
MDL Number MFCD00665911
InChI Key SEAVIOKUWQLSPU-UHFFFAOYSA-N
Synonym 2-amino-4-4-methylphenyl pyrimidine, 2-amino-4-p-tolyl pyrimidine, 4-p-tolyl pyrimidin-2-amine, 4-4-methylphenyl pyrimidin-2-amine, 4-4-methylphenyl pyrimidine-2-ylamine, acmc-20ao56, 2-pyrimidinamine, 4-4-methylphenyl
PubChem CID 2735370
IUPAC Name 4-(4-methylphenyl)pyrimidin-2-amine
SMILES CC1=CC=C(C=C1)C2=NC(=NC=C2)N

Specifications

Quantity 250 mg
Formula Weight 185.23
Percent Purity 96%
Chemical Name or Material 2-Amino-4-(4-methylphenyl)pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.