missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5,6,7,8-pentahydrothiazolo[5,4-d]azepine, TRC

Product Code. 30322662
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30322662 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30322662

Brand: Toronto Research Chemicals TRCA16893510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-4,5,6,7,8-pentahydrothiazolo[5,4-d]azepine
CAS 86029-68-7
Molecular Formula C7 H11 N3 S
Quantity 10 mg
InChI Formula InChI=1S/C7H11N3S/c8-7-10-5-1-3-9-4-2-6(5)11-7/h9H,1-4H2,(H2,8,10)
SMILES Nc1nc2CCNCCc2s1
IUPAC Name 5,6,7,8-tetrahydro-4H-[1,3]thiazolo[4,5-d]azepin-2-amine
Molecular Weight (g/mol) 169.25
Formula Weight 169.0674
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.