missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dihydroxypyrimidine, 98%

Product Code. 11469403
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469403 100 g 100g
11459403 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469403 Supplier Thermo Scientific Alfa Aesar Supplier No. A11813.22
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 56-09-7 | C4H5N3O2 | 127.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Amino-4,6-dihydroxypyrimidine acts as an intermediate in the production of antimicrobial guanylsulfonamides. It also acts as an intermediate in pharmaceuticals and agrochemicals.

Solubility
Soluble in Aqueous Alkali.

Notes
Store away from oxidizing agents. Store at -20°C. Store in a cool, dry, well-ventilated area.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-09-7
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006094
InChI Key AUFJTVGCSJNQIF-UHFFFAOYSA-N
Synonym 2-amino-4,6-dihydroxypyrimidine, 2-aminopyrimidine-4,6-diol, 4 1h-pyrimidinone, 2-amino-6-hydroxy, 2-amino-6-hydroxypyrimidin-4 3h-one, 2-amino-4,6-dioxypyrimidine, 2-amino-4,6-pyrimidinedione, 4,6-dihydroxy-2-aminopyrimidine, 2-amino-4,6-pyrimidinediol, 2-amino-6-hydroxy-1h-pyrimidin-4-one, 2-amino-6-hydroxy-4 1h-pyrimidinone
PubChem CID 66131
IUPAC Name 2-amino-4-hydroxy-1H-pyrimidin-6-one
SMILES NC1=NC(O)=CC(=O)N1

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 510297
Solubility Information Soluble in Aqueous Alkali.
Formula Weight 127.1
Percent Purity 98%
Chemical Name or Material 2-Amino-4,6-dihydroxypyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.