missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dihydroxypyrimidine, 98%

Product Code. 10284600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10284600 25 g Glass bottle 25g
10142270 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10284600 Supplier Thermo Scientific Acros Supplier No. 103510250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-09-7 | C4H5N3O2 | 127.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-09-7
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006094
InChI Key AUFJTVGCSJNQIF-UHFFFAOYSA-N
Synonym 2-amino-4,6-dihydroxypyrimidine, 2-aminopyrimidine-4,6-diol, 4 1h-pyrimidinone, 2-amino-6-hydroxy, 2-amino-6-hydroxypyrimidin-4 3h-one, 2-amino-4,6-dioxypyrimidine, 2-amino-4,6-pyrimidinedione, 4,6-dihydroxy-2-aminopyrimidine, 2-amino-4,6-pyrimidinediol, 2-amino-6-hydroxy-1h-pyrimidin-4-one, 2-amino-6-hydroxy-4 1h-pyrimidinone
PubChem CID 66131
IUPAC Name 2-amino-4-hydroxy-1H-pyrimidin-6-one
SMILES NC1=NC(O)=CC(=O)N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Pink to White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 24, 468
Formula Weight 127.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-4, 6-dihydroxypyrimidine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.