missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chloropyrimidine, 95%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| CAS | 3993-78-0 |
| Melting Point | 155°C to 160°C |
| Assay Percent Range | 94% min. (HPLC) |
| Packaging | Glass Bottle |
| Molecular Formula | C4H4ClN3 |
| Quantity | 1 g |
| Synonym | 2-amino-4-chloropyrimidine, 2-amine-4-chloropyrimidine, 2-pyrimidinamine, 4-chloro, 4-chloro-2-pyrimidinamine, 4-chloro-2-aminopyrimidine, 4-chloropyrimidine-2-amine, 4-chloro-pyrimidin-2-ylamine, 4-chloropyrimidine-2-ylamine, 2-amino-6-chloropyrimidine, 4-chloro-2-pyrimidineamine |
| InChI Key | DBGFGNCFYUNXLD-UHFFFAOYSA-N |
| SMILES | C1=CN=C(N=C1Cl)N |
| IUPAC Name | 4-chloropyrimidin-2-amine |
| Show More |
Safety and Handling
Product Identifier
- 2-Amino-4-chloropyrimidine, 95%
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction