missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylpyrimidine, TRC

Product Code. 30321206
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30321206 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30321206

Brand: Toronto Research Chemicals TRCA61623010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-4-methylpyrimidine
CAS 108-52-1
Molecular Formula C5H7N3
Quantity 10 g
Synonym 4-Methyl-2-pyrimidinamine, 2-Amino-6-methylpyrimidine, 4-Methyl-2-aminopyrimidine, 6-Methyl-2-pyrimidinamine, NSC 1939
InChI Formula InChI=1S/C5H7N3/c1-4-2-3-7-5(6)8-4/h2-3H,1H3,(H2,6,7,8)
SMILES Cc1ccnc(N)n1
IUPAC Name 4-methylpyrimidin-2-amine
Molecular Weight (g/mol) 109.13
Formula Weight 109.064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.