missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(p-tolyl)thiazole, 98%

Product Code. 11363287
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363287 5 g 5g
11353287 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363287 Supplier Thermo Scientific Alfa Aesar Supplier No. L13863.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2103-91-5 | C10H10N2S | 190.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2103-91-5
Molecular Formula C10H10N2S
Molecular Weight (g/mol) 190.264
MDL Number MFCD00170264
InChI Key ARLHWYFAPHJCJT-UHFFFAOYSA-N
Synonym 4-4-methylphenyl-1,3-thiazol-2-amine, 4-p-tolyl thiazol-2-amine, 2-amino-4-p-tolyl thiazole, 4-p-tolyl-thiazol-2-ylamine, 4-p-tolyl thiazol-2-ylamine, 4-4-methylphenyl-2-thiazolamine, 4-p-tolylthiazol-2-amine, 4-4-methylphenyl-1,3-thiazole-2-ylamine, 2-amino-4-4-methylphenyl thiazole, 4-4-methylphenyl-1,3-thiazol-2-ylamine
PubChem CID 244066
IUPAC Name 4-(4-methylphenyl)-1,3-thiazol-2-amine
SMILES CC1=CC=C(C=C1)C2=CSC(=N2)N

Specifications

Melting Point 134°C to 138°C
Quantity 5 g
Beilstein 143563
Formula Weight 190.27
Percent Purity 98%
Chemical Name or Material 2-Amino-4-(p-tolyl)thiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.