missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-phenylthiazole, 97%

Product Code. 10803771 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10803771 1 g Glass bottle 1g
10412590 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10803771 Supplier Thermo Scientific Acros Supplier No. 432490010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2010-06-2 | C9H8N2S | 176.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2010-06-2
Molecular Formula C9H8N2S
Molecular Weight (g/mol) 176.24
MDL Number MFCD00039680
InChI Key PYSJLPAOBIGQPK-UHFFFAOYSA-N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 150.0°C to 152.5°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 4-phenylthiazol-2-amine, 2-amino-4-phenylthiazole, phenthiazamine, 2-thiazolamine, 4-phenyl, 4-phenyl-2-thiazolamine, thiazole, 2-amino-4-phenyl, 4-phenyl-thiazol-2-ylamine, 4-phenyl-2-thiazolylamine, 2-amino-4-phenyl-1,3-thiazole, 4-phenyl-2-aminothiazole
SMILES C1=CC=C(C=C1)C2=CSC(=N2)N
IUPAC Name 4-phenyl-1,3-thiazol-2-amine
PubChem CID 40302
Formula Weight 176.24
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 2-Amino-4-phenylthiazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-4-phenylthiazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.