missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-thiazoleacetic acid, 97%

Product Code. 11445547
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445547 100 g 100g
11435547 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445547 Supplier Thermo Scientific Alfa Aesar Supplier No. B23534.22
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29676-71-9 | C5H6N2O2S | 158.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29676-71-9
Molecular Formula C5H6N2O2S
Molecular Weight (g/mol) 158.175
MDL Number MFCD00010068
InChI Key DYCLHZPOADTVKK-UHFFFAOYSA-N
Synonym 2-amino-4-thiazoleacetic acid, 2-2-aminothiazol-4-yl acetic acid, 2-aminothiazol-4-acetic acid, 2-aminothiazole-4-yl acetic acid, 4-thiazoleacetic acid, 2-amino, 2-amino-1,3-thiazol-4-yl acetic acid, 2-aminothiazole-4-yl-acetic acid, 2-aminothiazole-4-acetic acid, unii-qsh3o4tnjt, 2-2-amino-1,3-thiazol-4-yl acetic acid
PubChem CID 34665
IUPAC Name 2-(2-amino-1,3-thiazol-4-yl)acetic acid
SMILES C1=C(N=C(S1)N)CC(=O)O

Specifications

Melting Point ∼130°C (decomposition)
Quantity 100 g
Beilstein 127415
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material 2-Amino-4-thiazoleacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.