missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-thiazoleacetic Acid, TRC

Product Code. 30134678
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134678 50 g 50g
30132966 100 g 100g
30006150 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134678 Supplier Toronto Research Chemicals Supplier No. TRCA63048550G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29676-71-9
Molecular Formula C5H6N2O2S
Molecular Weight (g/mol) 158.18
Synonym 2-Amino-4-thiazoleacetic Acid, (2-Amino-1,3-thiazol-4-yl)acetic acid, (2-Amino-4-thiazoyl)acetic acid, 2-(2-Amino-4-thiazole)acetic acid, 2-(2-Amino-4-thiazolyl)acetic acid, 2-Amino-4-thiazolylacetic acid, 2-Aminothiazol-4-acetic acid, 2-Aminothiazole-4-acetic acid, SQ 272
IUPAC Name 2-(2-amino-1,3-thiazol-4-yl)acetic acid
SMILES Nc1nc(CC(=O)O)cs1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C5H6N2O2S/c6-5-7-3(2-10-5)1-4(8)9/h2H,1H2,(H2,6,7)(H,8,9)
Formula Weight 158.015
Chemical Name or Material 2-Aminothiazole-4-acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.